Learn More
TARGETMOL CHEMICALS INC CUCURBITACIN E 5MG
SDP
Catalog No. 502576638
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cucurbitacin E is a natural product isolated from the climbing stem of Cucumic melo L. It significantly suppresses the activity of the cyclin B1/CDC2 complex.Cucurbitacin E has prevention of neurodegeneration it has potent anti-proliferative antineoplastic anti-inflammatory and analgesic actions. purity: 98%
Catalog No. 50-257-6638 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S14675MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Cucurbitacin E is a natural product isolated from the climbing stem of Cucumic melo L. It significantly suppresses the activity of the cyclin B1/CDC2 complex.Cucurbitacin E has prevention of neurodegeneration it has potent anti-proliferative antineoplastic anti-inflammatory and analgesic actions. purity: 98%