Learn More
Cayman Chemical CucurbItacIn I 10mg
SDP
Catalog No. 502972647
Encompass_Preferred
A potent inhibitor of the STAT3/JAK signaling pathway; suppresses levels of tyrosine phosphorylated STAT3, STAT3 DNA binding and STAT3-mediated gene transcription; suppresses tumor growth in vivo
Catalog No. 50-297-2647 Supplier Cayman Chemical Supplier No. 1474710 MG
A potent inhibitor of the STAT3/JAK signaling pathway; suppresses levels of tyrosine phosphorylated STAT3, STAT3 DNA binding and STAT3-mediated gene transcription; suppresses tumor growth in vivo