Learn More
Cayman Chemical Cucurbitacin IIA 10MG
SDP
Catalog No. 502915252
Encompass_Preferred
A triterpenoid with diverse biological activities; decreases the proliferation of HT-29 cells (IC50 = 370 nM); induces cell cycle arrest at the G2/M phase and apoptosis in HT-29 cells at 1 µM; induces cell cycle arrest at the G0/G1 phase in LPS-stimulated, but not unstimulated, RAW 264.7 macrophages at 2.5 µM; decreases immobility time in the forced swim test and the tail suspension test in a mouse model of intermittent unpredictable stress at 5 mg/kg
Catalog No. 50-291-5252 Supplier Cayman Chemical Supplier No. 3737110 MG
A triterpenoid with diverse biological activities; decreases the proliferation of HT-29 cells (IC50 = 370 nM); induces cell cycle arrest at the G2/M phase and apoptosis in HT-29 cells at 1 µM; induces cell cycle arrest at the G0/G1 phase in LPS-stimulated, but not unstimulated, RAW 264.7 macrophages at 2.5 µM; decreases immobility time in the forced swim test and the tail suspension test in a mouse model of intermittent unpredictable stress at 5 mg/kg