Learn More
TARGETMOL CHEMICALS INC Cucurbitacin IIA 10MG
SDP
Catalog No. 502878816
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 1. Cucurbitacin IIA (Dihydrocucurbitacin Q1) can induce apoptosis and enhance autophagy , contributes to the anti-inflammatory activity of Cucurbitacin IIA against inflammation-related diseases. 2. Cucurbitacin IIA is a novel class of anti-cancer drug in suppression of cancer cell expansion by disrupting the actin cytoskeleton and directing the cell to undergo PARP-mediated apoptosis through the inhibition of survivin downstream of JAK2/STAT3. Purity 98.62%
Catalog No. 50-287-8816 Supplier TARGETMOL CHEMICALS INC Supplier No. T3S147110MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. 1. Cucurbitacin IIA (Dihydrocucurbitacin Q1) can induce apoptosis and enhance autophagy , contributes to the anti-inflammatory activity of Cucurbitacin IIA against inflammation-related diseases. 2. Cucurbitacin IIA is a novel class of anti-cancer drug in suppression of cancer cell expansion by disrupting the actin cytoskeleton and directing the cell to undergo PARP-mediated apoptosis through the inhibition of survivin downstream of JAK2/STAT3. Purity 98.62%