Learn More
Apexbio Technology LLC CUCURBITACIN-IIA-20MG
SDP
Catalog No. 5000567428
Encompass_Preferred
Cucurbitacin a (CAS No 58546-34-2) is a significant compound in biomedical research primarily studied for its potential anticancer properties Originating from the Cucurbitaceae plant family its mechanism of action involves targeting multiple cellular pathways including JAK/STAT and MAPK leading to the inhibition of cancer cell proliferation and induction of apoptosis It exhibits activity predominantly in the nanomolar to low micromolar range Commonly utilized in various in vitro and in vivo models Cucurbitacin a is tested in numerous cancer cell lines and animal studies to evaluate its efficacy and potential as a chemotherapeutic agent Its application extends to drug screening processes where experimental concentrations typically depend on the specific research design
Catalog No. 50-005-67428 Supplier Apexbio Technology LLC Supplier No. N220120
Cucurbitacin a (CAS No 58546-34-2) is a significant compound in biomedical research primarily studied for its potential anticancer properties Originating from the Cucurbitaceae plant family its mechanism of action involves targeting multiple cellular pathways including JAK/STAT and MAPK leading to the inhibition of cancer cell proliferation and induction of apoptosis It exhibits activity predominantly in the nanomolar to low micromolar range Commonly utilized in various in vitro and in vivo models Cucurbitacin a is tested in numerous cancer cell lines and animal studies to evaluate its efficacy and potential as a chemotherapeutic agent Its application extends to drug screening processes where experimental concentrations typically depend on the specific research design