Learn More
Medchemexpress LLC Cucurbitacin IIa | 58546-34-2 | 562.73 | 1 MG
SDP
Catalog No. 5000414682
Encompass_Preferred
Cucurbitacin IIa, also known as Hemslecin A, is an orally active EGFR inhibitor capable of permeating the blood-brain barrier. It demonstrates an IC50 of 1.455 nM against human EGFR and exhibits anti-inflammatory activity by inhibiting the EGFR-MAPK signaling pathway.
Features and Benefits:
  • Induces caspase-3-dependent apoptosis.
  • Downregulates survivin expression.
  • Enhances autophagy levels.
  • Disrupts the actin cytoskeleton via actin aggregation.
  • Arrests the cell cycle at the G2/M phase.
  • Supports research into inflammation-related diseases.
  • Useful in studies on depression and cancers like non-small cell lung cancer.
Catalog No. 50-004-14682 Supplier Medchemexpress LLC Supplier No. HYN19881MG
Cucurbitacin IIa, also known as Hemslecin A, is an orally active EGFR inhibitor capable of permeating the blood-brain barrier. It demonstrates an IC50 of 1.455 nM against human EGFR and exhibits anti-inflammatory activity by inhibiting the EGFR-MAPK signaling pathway.
Features and Benefits:
  • Induces caspase-3-dependent apoptosis.
  • Downregulates survivin expression.
  • Enhances autophagy levels.
  • Disrupts the actin cytoskeleton via actin aggregation.
  • Arrests the cell cycle at the G2/M phase.
  • Supports research into inflammation-related diseases.
  • Useful in studies on depression and cancers like non-small cell lung cancer.