Learn More
Medchemexpress LLC Cucurbitacin IIa | 58546-34-2 | 562.73 | 1 ML
SDP
Catalog No. 5000414591
Encompass_Preferred
Cucurbitacin IIa, also known as Hemslecin A, is an orally active and blood-brain barrier-permeable EGFR inhibitor, demonstrating an IC50 of 1.455 nM against human EGFR. It plays a role in inducing caspase-3-dependent apoptosis, downregulating survivin expression, and enhancing autophagy levels. This compound disrupts the actin cytoskeleton through actin aggregation, arrests the cell cycle at the G2/M phase, and provides anti-inflammatory effects by inhibiting the EGFR-MAPK signaling pathway.
  • Orally active, blood-brain barrier-permeable EGFR inhibitor
  • Induces caspase-3-dependent apoptosis
  • Downregulates survivin expression
  • Enhances autophagy levels
  • Disrupts actin cytoskeleton via actin aggregation
  • Arrests cell cycle at G2/M phase
  • Exerts anti-inflammatory activity by inhibiting EGFR-MAPK signaling pathway
  • Suitable for research in inflammation-related diseases, depression, and cancers
Catalog No. 50-004-14591 Supplier Medchemexpress LLC Supplier No. HYN198810MM/1ML
Cucurbitacin IIa, also known as Hemslecin A, is an orally active and blood-brain barrier-permeable EGFR inhibitor, demonstrating an IC50 of 1.455 nM against human EGFR. It plays a role in inducing caspase-3-dependent apoptosis, downregulating survivin expression, and enhancing autophagy levels. This compound disrupts the actin cytoskeleton through actin aggregation, arrests the cell cycle at the G2/M phase, and provides anti-inflammatory effects by inhibiting the EGFR-MAPK signaling pathway.
  • Orally active, blood-brain barrier-permeable EGFR inhibitor
  • Induces caspase-3-dependent apoptosis
  • Downregulates survivin expression
  • Enhances autophagy levels
  • Disrupts actin cytoskeleton via actin aggregation
  • Arrests cell cycle at G2/M phase
  • Exerts anti-inflammatory activity by inhibiting EGFR-MAPK signaling pathway
  • Suitable for research in inflammation-related diseases, depression, and cancers