Learn More
Medchemexpress LLC HY-N1987 5mg Medchemexpress, Cucurbitacin IIb CAS:50298-90-3 Purity:>98%
SDP
Catalog No. 501970355
Encompass_Preferred
Medchemexpress, HY-N1987 5mg Cucurbitacin IIb CAS:50298-90-3 Cucurbitacin IIb is an active component isolated from Hemsleya amabilis, induces apoptosis with anti-inflammatory activity. Cucurbitacin IIb inhibits phosphorylation of STAT3, JNK and Erk1/2, enhances the phosphorylation of IκB and NF-κB (p65), blocks nuclear translocation of NF-κB (p65) and decreases mRNA levels of IκBα and TNF-α. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0355 Supplier Medchemexpress LLC Supplier No. HYN19875MG
Medchemexpress, HY-N1987 5mg Cucurbitacin IIb CAS:50298-90-3 Cucurbitacin IIb is an active component isolated from Hemsleya amabilis, induces apoptosis with anti-inflammatory activity. Cucurbitacin IIb inhibits phosphorylation of STAT3, JNK and Erk1/2, enhances the phosphorylation of IκB and NF-κB (p65), blocks nuclear translocation of NF-κB (p65) and decreases mRNA levels of IκBα and TNF-α. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.