Learn More
Medchemexpress LLC Cucurbitacin Q1 | 99530-82-2 | 5 MG
SDP
Catalog No. 5000417353
Encompass_Preferred
Cucurbitacin Q1 is a tetracyclic triterpene, isolated from *Cucumis prophetarum*. It is a white to off-white solid with a molecular weight of 560.72 and chemical formula C32H48O8. Cucurbitacin Q1 exhibits potent cytotoxicity against various human cancer cell lines, including HT-29 (IC50: 0.54 μM), MCF7 (GI50: 0.11 μM), NCI-H460 (GI50: 0.065 μM), and SF-268 (GI50: 0.087 μM), indicating its potential in anti-cancer research. Its CAS number is 99530-82-2.
  • High purity: 98.39%.
  • Available in various package sizes.
  • Comprehensive documentation including data sheet, SDS, COA, and more.
  • Detailed protocols for dissolution in both in vitro and in vivo studies.
  • Recommended storage at 4°C, protected from light.
Catalog No. 50-004-17353 Supplier Medchemexpress LLC Supplier No. HYN81375MG
Cucurbitacin Q1 is a tetracyclic triterpene, isolated from *Cucumis prophetarum*. It is a white to off-white solid with a molecular weight of 560.72 and chemical formula C32H48O8. Cucurbitacin Q1 exhibits potent cytotoxicity against various human cancer cell lines, including HT-29 (IC50: 0.54 μM), MCF7 (GI50: 0.11 μM), NCI-H460 (GI50: 0.065 μM), and SF-268 (GI50: 0.087 μM), indicating its potential in anti-cancer research. Its CAS number is 99530-82-2.
  • High purity: 98.39%.
  • Available in various package sizes.
  • Comprehensive documentation including data sheet, SDS, COA, and more.
  • Detailed protocols for dissolution in both in vitro and in vivo studies.
  • Recommended storage at 4°C, protected from light.