Learn More
Medchemexpress LLC Cucurbituril | 80262-44-8 | MFCD10567435 | 97.0% | 996.82 | C36H36N24O12 | 250 MG
SDP
Catalog No. 5000282976
Encompass_Preferred
Cucurbituril is a macrocyclic "container" molecule used as a host in supramolecular chemistry and biochemical assays. It features a hydrophobic interior cavity and two carbonyl-lined portals that enable selective binding of guest molecules. Supplied as a solid for research applications.
  • Macrocyclic host molecule with a hydrophobic cavity for selective guest binding.
  • Useful for host-guest chemistry, molecular recognition, and catalysis studies.
  • Characterized by CAS 80262-44-8 and molecular formula C36H36N24O12.
  • Available in multiple package sizes, including 250 MG.
  • Purity typically confirmed by NMR (reported 97.0%).
Catalog No. 50-002-82976 Supplier Medchemexpress LLC Supplier No. HYW130354250MG
Cucurbituril is a macrocyclic "container" molecule used as a host in supramolecular chemistry and biochemical assays. It features a hydrophobic interior cavity and two carbonyl-lined portals that enable selective binding of guest molecules. Supplied as a solid for research applications.
  • Macrocyclic host molecule with a hydrophobic cavity for selective guest binding.
  • Useful for host-guest chemistry, molecular recognition, and catalysis studies.
  • Characterized by CAS 80262-44-8 and molecular formula C36H36N24O12.
  • Available in multiple package sizes, including 250 MG.
  • Purity typically confirmed by NMR (reported 97.0%).