Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cuda | 479413-68-8 | 99.8% | 5 MG
SDP

Catalog No. 5000231106
Encompass_Preferred

Cuda is a small-molecule research reagent that functions as a potent inhibitor of soluble epoxide hydrolase (sEH) and a modulator of PPARα activity. It is supplied as a solid for laboratory research and has reported IC50 values of 11.1 nM (mouse sEH) and 112 nM (human sEH).

  • Potent sEH inhibitor with low-nanomolar activity in mouse
  • Modulates PPARα activity in cellular assays
  • High purity (99.8% as provided)
  • Available in small-mass quantities for screening and assay development
  • Store powder at -20°C for long-term stability

Catalog No. 50-002-31106 Supplier Medchemexpress LLC Supplier No. HY1215385MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cuda is a small-molecule research reagent that functions as a potent inhibitor of soluble epoxide hydrolase (sEH) and a modulator of PPARα activity. It is supplied as a solid for laboratory research and has reported IC50 values of 11.1 nM (mouse sEH) and 112 nM (human sEH).

  • Potent sEH inhibitor with low-nanomolar activity in mouse
  • Modulates PPARα activity in cellular assays
  • High purity (99.8% as provided)
  • Available in small-mass quantities for screening and assay development
  • Store powder at -20°C for long-term stability

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.