Learn More
Medchemexpress LLC Cuda disodium | 98.0% | 386.48 g/mol | C19H36N2Na2O3 | 5 MG
SDP
Catalog No. 5000264833
Encompass_Preferred
CUDA disodium is a potent, research-use small molecule inhibitor of soluble epoxide hydrolase (sEH) that modulates PPARα activity. Reported IC50 values are 11.1 nM (mouse sEH) and 112 nM (human sEH). Supplied as a 5 mg sample with 98.0% purity, it is intended for biochemical and pharmacological research in lipid metabolism and cardiovascular disease models.
  • Potent sEH inhibition with reported IC50s of 11.1 nM (mouse) and 112 nM (human).
  • Enhances peroxisome proliferator-activated receptor alpha activity in cellular models.
  • High purity (98.0%) suitable for biochemical assays.
  • Small quantity packaging (5 mg) for screening and early-stage studies.
  • Storage recommendations provided for stability in solvent and dry conditions.
Catalog No. 50-002-64833 Supplier Medchemexpress LLC Supplier No. HY121538A5MG
CUDA disodium is a potent, research-use small molecule inhibitor of soluble epoxide hydrolase (sEH) that modulates PPARα activity. Reported IC50 values are 11.1 nM (mouse sEH) and 112 nM (human sEH). Supplied as a 5 mg sample with 98.0% purity, it is intended for biochemical and pharmacological research in lipid metabolism and cardiovascular disease models.
  • Potent sEH inhibition with reported IC50s of 11.1 nM (mouse) and 112 nM (human).
  • Enhances peroxisome proliferator-activated receptor alpha activity in cellular models.
  • High purity (98.0%) suitable for biochemical assays.
  • Small quantity packaging (5 mg) for screening and early-stage studies.
  • Storage recommendations provided for stability in solvent and dry conditions.