Learn More
Selleck Chemical LLC CUDC-101 S1194-10mM/1mL
SDP
Catalog No. 503111443
Encompass_Preferred
CUDC-101 is a potent multi-targeted inhibitor against HDAC EGFR and HER2 with IC50 of 4 4 nM 2 4 nM and 15 7 nM and inhibits class I/II HDACs but not class III Sir-type HDACs Phase 1
Catalog No. 50-311-1443 Supplier Selleck Chemical LLC Supplier No. S119410MM/1ML
CUDC-101 is a potent multi-targeted inhibitor against HDAC EGFR and HER2 with IC50 of 4 4 nM 2 4 nM and 15 7 nM and inhibits class I/II HDACs but not class III Sir-type HDACs Phase 1