Learn More
Apexbio Technology LLC CUDC-101 1012054-59-9 50mg
SDP
Catalog No. 5000064225
Encompass_Preferred
CUDC-101 (CAS 1012054-59-9) is a multitargeted small molecule inhibitor that concurrently targets epidermal growth factor receptor (EGFR) HER2 and class I and II histone deacetylases (HDACs) By inhibiting EGFR and HER2 kinases directly and simultaneously blocking HDAC activity CUDC-101 downregulates multiple oncogenic signaling pathways including those mediated by Akt HER3 and MET Preclinical assessments demonstrate that CUDC-101 inhibits growth of diverse tumor cell lines and xenograft models notably including those resistant to EGFR-targeted agents such as lapatinib and erlotinib These characteristics render CUDC-101 valuable for oncology research and drug resistance studies
Catalog No. 50-000-64225 Supplier Apexbio Technology LLC Supplier No. A409250
CUDC-101 (CAS 1012054-59-9) is a multitargeted small molecule inhibitor that concurrently targets epidermal growth factor receptor (EGFR) HER2 and class I and II histone deacetylases (HDACs) By inhibiting EGFR and HER2 kinases directly and simultaneously blocking HDAC activity CUDC-101 downregulates multiple oncogenic signaling pathways including those mediated by Akt HER3 and MET Preclinical assessments demonstrate that CUDC-101 inhibits growth of diverse tumor cell lines and xenograft models notably including those resistant to EGFR-targeted agents such as lapatinib and erlotinib These characteristics render CUDC-101 valuable for oncology research and drug resistance studies