Learn More
Medchemexpress LLC HY-10223 5mg Medchemexpress, CUDC-101 CAS:1012054-59-9 Purity:>98%
SDP
Catalog No. 501932230
Encompass_Preferred
Medchemexpress, HY-10223 5mg CUDC-101 CAS:1012054-59-9 CUDC-101 is a potent inhibitor of HDAC , EGFR , and HER2 with IC 50 s of 4.4, 2.4, and 15.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2230 Supplier Medchemexpress LLC Supplier No. HY102235MG
Medchemexpress, HY-10223 5mg CUDC-101 CAS:1012054-59-9 CUDC-101 is a potent inhibitor of HDAC , EGFR , and HER2 with IC 50 s of 4.4, 2.4, and 15.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.