Learn More
Selleck Chemical LLC CUDC-101 10mg 1012054-59-9
SDP
Catalog No. 508455
Encompass_Preferred
CUDC-101 is a potent multi-targeted inhibitor against HDAC, EGFR and HER2 with IC50 of 4.4 nM, 2.4 nM, and 15.7 nM, and inhibits class I/II HDACs, but not class III, Sir-type HDACs. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-845-5 Supplier Selleck Chemical LLC Supplier No. S119410MG
CUDC-101 is a potent multi-targeted inhibitor against HDAC, EGFR and HER2 with IC50 of 4.4 nM, 2.4 nM, and 15.7 nM, and inhibits class I/II HDACs, but not class III, Sir-type HDACs. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.