Learn More
TARGETMOL CHEMICALS INC CUDC-427 5MG
SDP
Catalog No. 502581259
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CUDC-427 (GDC-0917) is an orally bioactive and pan antagonist of inhibitor of apoptosis proteins(IAPs). CUDC-427 can be used in research on cancers. purity: 98%
Catalog No. 50-258-1259 Supplier TARGETMOL CHEMICALS INC Supplier No. T150185MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. CUDC-427 (GDC-0917) is an orally bioactive and pan antagonist of inhibitor of apoptosis proteins(IAPs). CUDC-427 can be used in research on cancers. purity: 98%