Learn More
Medchemexpress LLC HY-13522 50mg Medchemexpress, CUDC-907 fimepinostat CAS:1339928-25-4 Purity:98%
SDP
Catalog No. 501637966
Encompass_Preferred
Medchemexpress,HY-13522 50mg CUDC-907 fimepinostat CAS:1339928-25-4 Formula:C23H24N8O4S CUDC-907 potently inhibits class I PI3Ks as well as classes I and II HDAC enzymes with IC50 of 19/54/39 nM and 1.7/5.0/1.8/2.8 nM for PI3Kα/PI3Kβ/PI3Kδ and HDAC1/HDAC2/HDAC3/HDAC10 , respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-7966 Supplier Medchemexpress LLC Supplier No. HY1352250MG
Medchemexpress,HY-13522 50mg CUDC-907 fimepinostat CAS:1339928-25-4 Formula:C23H24N8O4S CUDC-907 potently inhibits class I PI3Ks as well as classes I and II HDAC enzymes with IC50 of 19/54/39 nM and 1.7/5.0/1.8/2.8 nM for PI3Kα/PI3Kβ/PI3Kδ and HDAC1/HDAC2/HDAC3/HDAC10 , respectively. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.