Learn More
Apexbio Technology LLC CUDC-907 1339928-25-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000061432
Encompass_Preferred
CUDC-907 (CAS 1339928-25-4) is a dual inhibitor targeting histone deacetylases (HDACs 1 2 3 10) and class I phosphatidylinositol 3-kinases (PI3K) It exhibits nanomolar inhibitory activity against these enzymes blocking PI3K-mediated Akt signaling and increasing acetylation of histones and non-histone substrates (e g tubulin p53) In cancer cell models CUDC-907 induces G2-M phase arrest and reduces phosphorylation of downstream proteins such as p70S6 4EBP-1 MEK STAT3 and SRC Preclinical studies demonstrate its anti-tumor activity in lymphoma and NSCLC xenografts making it relevant for oncology research
Catalog No. 50-000-61432 Supplier Apexbio Technology LLC Supplier No. A40975.1
CUDC-907 (CAS 1339928-25-4) is a dual inhibitor targeting histone deacetylases (HDACs 1 2 3 10) and class I phosphatidylinositol 3-kinases (PI3K) It exhibits nanomolar inhibitory activity against these enzymes blocking PI3K-mediated Akt signaling and increasing acetylation of histones and non-histone substrates (e g tubulin p53) In cancer cell models CUDC-907 induces G2-M phase arrest and reduces phosphorylation of downstream proteins such as p70S6 4EBP-1 MEK STAT3 and SRC Preclinical studies demonstrate its anti-tumor activity in lymphoma and NSCLC xenografts making it relevant for oncology research