Learn More
Cayman Chemical Cudetaxestat, Purity: ≥98%, CAS: 1782070-21-6, 10 mg
SDP
Catalog No. 502917609
Encompass_Preferred
An autotaxin inhibitor (IC50 = ≤0.5 µM); reduces MDA-MB-435 cell migration (IC50 = 15 nM); reduces blood glucose levels in a glucose tolerance test, as well as baseline blood glucose levels, in mice fed a high-fat diet at 15 mg/kg twice per day for two days; reduces liver fibrosis in a CDAA/HFD mouse model of liver fibrosis at 10 mg/kg; reduces colon ulcer area and increases colon length in mouse models of acute and chronic ulcerative colitis, respectively
Catalog No. 50-291-7609 Supplier Cayman Chemical Supplier No. 3878910 MG
An autotaxin inhibitor (IC50 = ≤0.5 µM); reduces MDA-MB-435 cell migration (IC50 = 15 nM); reduces blood glucose levels in a glucose tolerance test, as well as baseline blood glucose levels, in mice fed a high-fat diet at 15 mg/kg twice per day for two days; reduces liver fibrosis in a CDAA/HFD mouse model of liver fibrosis at 10 mg/kg; reduces colon ulcer area and increases colon length in mouse models of acute and chronic ulcerative colitis, respectively