Learn More
FENWAL INC CUE PRIM SET 4/CS
Catalog No. NC2605834
Encompass
  • Single-use design for precise small-volume cell processing.
  • 4 µm filter pore size for effective separation.
  • Integrates syringe pumps and valving cassette for high accuracy.
  • Supports concentration, washing, dilution, formulation, and aliquoting of cells.
  • Configurable protocols for consistent processing.
  • Functionally closed system for enhanced safety.
  • Designed for laboratory use only.
  • Filtrate bag volume of 2.5 L.
  • Includes 3 individually packaged sampling sets.
  • Two syringe assemblies come with integrated 0.2 µm air filters.
  • Cassette includes syringe holders and a 0.2 µm air filter.
  • Tubing includes source line and 3 solution lines for versatility.
Catalog No. NC2605834 Supplier FENWAL INC Supplier No. X6R5004
Add to Cart
Add to Cart
Request a Quote
  • Single-use design for precise small-volume cell processing.
  • 4 µm filter pore size for effective separation.
  • Integrates syringe pumps and valving cassette for high accuracy.
  • Supports concentration, washing, dilution, formulation, and aliquoting of cells.
  • Configurable protocols for consistent processing.
  • Functionally closed system for enhanced safety.
  • Designed for laboratory use only.
  • Filtrate bag volume of 2.5 L.
  • Includes 3 individually packaged sampling sets.
  • Two syringe assemblies come with integrated 0.2 µm air filters.
  • Cassette includes syringe holders and a 0.2 µm air filter.
  • Tubing includes source line and 3 solution lines for versatility.