Learn More
EUROTROL CUESEE VERISTAT LEVEL 1 10/BOX
Catalog No. NC0545046
Encompass
For use with the Abbott i-STAT analyzer to verify precision and accuracy of all blood gases, electrolyes, metabolite and hematocrit. Added blue dye to assist in cartridge dosing. Shelf life of 2 years, room temperature stability of 10 days. Availabl
Catalog No. NC0545046 Supplier EUROTROL Supplier No. 221.001.010
For use with the Abbott i-STAT analyzer to verify precision and accuracy of all blood gases, electrolyes, metabolite and hematocrit. Added blue dye to assist in cartridge dosing. Shelf life of 2 years, room temperature stability of 10 days. Availabl