Learn More
EUROTROL CUESEE VERISTAT LEVEL 3 10/BOX
Catalog No. NC0545048
Encompass
For use with the Abbott i-STAT analyzer to verify precision and accuracy of all blood gases, electrolyes, metabolite and hematocrit. Added blue dye to assist in cartridge dosing. Shelf life of 2 years, room temperature stability of 10 days. Availabl
Catalog No. NC0545048 Supplier EUROTROL Supplier No. 221.003.010
For use with the Abbott i-STAT analyzer to verify precision and accuracy of all blood gases, electrolyes, metabolite and hematocrit. Added blue dye to assist in cartridge dosing. Shelf life of 2 years, room temperature stability of 10 days. Availabl