Learn More
EUROTROL CUESEE VERISTAT LVM, ALL 5 LVS
Catalog No. NC0533160
Encompass
For use with the Abbott i-STAT analyzer to verify precision and accuracy of all blood gases, electrolyes, metabolite and hematocrit. Added blue dye to assist in cartridge dosing. Shelf life of 2 years, room temperature stability of 10 days. 5 clinic
Catalog No. NC0533160 Supplier EUROTROL Supplier No. 220.000.005
For use with the Abbott i-STAT analyzer to verify precision and accuracy of all blood gases, electrolyes, metabolite and hematocrit. Added blue dye to assist in cartridge dosing. Shelf life of 2 years, room temperature stability of 10 days. 5 clinic