Learn More
Enterprise Technology Solutions ADC Adcuff Cuff and Bladder with Two Tubes - Small Adult
SDP
Catalog No. 502407276
Encompass_Preferred
The ADC Adcuff Cuff and Bladder with Two Tubes - Small Adult is designed for use with most traditional pocket, clock, or mercury style aneroid instruments, and many blood pressure monitors, and includes an Adcuff Nylon Cuff and an inflation bladder with two tubes.
Catalog No. 50-240-7276 Supplier Enterprise Technology Solutions Supplier No. ADC84510SAAD2
The ADC Adcuff Cuff and Bladder with Two Tubes - Small Adult is designed for use with most traditional pocket, clock, or mercury style aneroid instruments, and many blood pressure monitors, and includes an Adcuff Nylon Cuff and an inflation bladder with two tubes.