Learn More
World Precision Instrument Cuff Liners for II-MRBP, 7/16", 100/PACK
SDP
Catalog No. 502538774
Encompass
WPI's Mouse and rat tail cuff blood pressure measuring systems for preclincial research  a true turn-key system when it is key critical for determing accurate, consistent blood pressure on mice, rats or any other laboratory animal test subject. A compact, simple yet versatile system that can test from a single up to 200 animals at a time with independent control of each channel. Simple daisy-chaining of systems allow expansion of systems, up to 200 independently, controlled channels. No other system in the world has such vast capabilities as the IITC blood pressure systems. Single animal systems are controlled from the touch screen which allows keying in all necessary test setups. These cuff liners are designed for use with the II-MRBP blood pressure system. Choose either 0.75" or 0.44" thick cuff liners. Both are sold in packages of 100 liners.
Catalog No. 50-253-8774 Supplier World Precision Instrument Supplier No. II2CL
WPI's Mouse and rat tail cuff blood pressure measuring systems for preclincial research  a true turn-key system when it is key critical for determing accurate, consistent blood pressure on mice, rats or any other laboratory animal test subject. A compact, simple yet versatile system that can test from a single up to 200 animals at a time with independent control of each channel. Simple daisy-chaining of systems allow expansion of systems, up to 200 independently, controlled channels. No other system in the world has such vast capabilities as the IITC blood pressure systems. Single animal systems are controlled from the touch screen which allows keying in all necessary test setups. These cuff liners are designed for use with the II-MRBP blood pressure system. Choose either 0.75" or 0.44" thick cuff liners. Both are sold in packages of 100 liners.