Learn More
Aviva Systems Biology Corp CUGBP1 Antibody - middle region (ARP40327_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40327_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CUGBP1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human CUGBP1. Alternate names: BRUNOL2, CUG-BP, CUGBP, CUGBP1, EDEN-BP, NAB50, NAPOR, hNab50ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57394