Learn More
Aviva Systems Biology Corp CUGBP2 Antibody - N-terminal region (ARP40667_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40667_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CUGBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CUGBP2. Alternate names: BRUNOL3, ETR-3, NAPOR, ETR3, CUGBP2ApplicationIHC, WB. Species reactivity: Cow, Human
Catalog No. 50-014-57580