Learn More
Aviva Systems Biology Corp CUL1 Antibody - C-terminal region (AVARP03049_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP03049_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against CUL1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human CUL1. Alternate names: ApplicationWB. Species reactivity: Cow, Dog, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-62529