Learn More
Sino Biological Recombinant Human CUL1/RBX1 Complex Protein (GST Tag)
Catalog No. 5000084239
Encompass_Preferred
Recombinant full-length human CUL1 and RBX1 were expressed by baculovirus in Sf9 insect cells using an N-terminal GST tags.
Catalog No. 50-000-84239 Supplier Sino Biological Supplier No. U27534G20
Recombinant full-length human CUL1 and RBX1 were expressed by baculovirus in Sf9 insect cells using an N-terminal GST tags.