Learn More
Aviva Systems Biology Corp CUL3 Antibody - C-terminal region (ARP61264_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61264_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CUL3. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human CUL3. Alternate names: CUL-3, PHA2EApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-316