Learn More
Aviva Systems Biology Corp CUL4A Antibody - middle region (ARP61665_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61665_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CUL4A. It was validated on Western Blot. . Alternate names: -ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-435