Learn More
Aviva Systems Biology Corp Cul4b Antibody - C-terminal region (ARP62057_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP62057_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Cul4b. It was validated on Western Blot. . Alternate names: 2700050M05Rik, AA409770, KIAA0695, mKIAA0695, CUL-4BApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-287-554