Learn More
Bioanalytical Systems Inc CULEX CHADS-VIOLET-192 PER SH
SDP
Catalog No. 50854145
Encompass
Tags, Chads; Attach to vials without the use of adhesives and stay in place due to friction; Waterproof; Solvent resistant; 10 sheets; 192 per sheet; Violet; 5.2 to 6.0 mm OD *Does not ship to Puerto Rico
Catalog No. 50-854-145 Supplier Bioanalytical Systems Inc Supplier No. CX1800V
Tags, Chads; Attach to vials without the use of adhesives and stay in place due to friction; Waterproof; Solvent resistant; 10 sheets; 192 per sheet; Violet; 5.2 to 6.0 mm OD *Does not ship to Puerto Rico