Learn More
Bioanalytical Systems Inc CULEX CHDS-BLUE-192 PER SHEET
SDP
Catalog No. 50854141
Encompass
Tags, Chads; Attach to vials without the use of adhesives and stay in place due to friction; Waterproof; Solvent resistant; 10 sheets; 192 per sheet; Blue; 5.2 to 6.0 mm OD *Does not ship to Puerto Rico
Catalog No. 50-854-141 Supplier Bioanalytical Systems Inc Supplier No. CX1800B
Tags, Chads; Attach to vials without the use of adhesives and stay in place due to friction; Waterproof; Solvent resistant; 10 sheets; 192 per sheet; Blue; 5.2 to 6.0 mm OD *Does not ship to Puerto Rico