Learn More
Bioanalytical Systems Inc CULEX CHDS-SALMON-192 PER SHT
SDP
Catalog No. 50854144
Encompass
Tags, Chads; Attach to vials without the use of adhesives and stay in place due to friction; Waterproof; Solvent resistant; 10 sheets; 192 per sheet; Salmon; 5.2 to 6.0 mm OD *Does not ship to Puerto Rico
Catalog No. 50-854-144 Supplier Bioanalytical Systems Inc Supplier No. CX1800S
Tags, Chads; Attach to vials without the use of adhesives and stay in place due to friction; Waterproof; Solvent resistant; 10 sheets; 192 per sheet; Salmon; 5.2 to 6.0 mm OD *Does not ship to Puerto Rico