Learn More
Bioanalytical Systems Inc CULEX EXTENDER -DRUG INFUSION
SDP
Catalog No. 50854176
Encompass
Extender, Catheter; Use to dose an animal manually from outside the cage; No bead; Dead volume: 22uL; Each *Does not ship to Puerto Rico
Catalog No. 50-854-176 Supplier Bioanalytical Systems Inc Supplier No. CX2030S
Extender, Catheter; Use to dose an animal manually from outside the cage; No bead; Dead volume: 22uL; Each *Does not ship to Puerto Rico