Learn More
Bioanalytical Systems Inc CULEX EXTENDER-GI W/LUER FTG
SDP
Catalog No. 50854173
Encompass
Extender, G.I. Catheter; Use to orally dose an animal manually from outside the cage; No bead; Dead volume: 186uL; Each *Does not ship to Puerto Rico
Catalog No. 50-854-173 Supplier Bioanalytical Systems Inc Supplier No. CX2029S
Extender, G.I. Catheter; Use to orally dose an animal manually from outside the cage; No bead; Dead volume: 186uL; Each *Does not ship to Puerto Rico