Learn More
Bioanalytical Systems Inc CULEX FEMORAL-SHORT-DK BLUE
SDP
Catalog No. 50854165
Encompass
Catheter, Short Femoral; Use to cannulate the femoral vein; Bead Color: Dark Blue; Dead Volume: 56uL; Each *Does not ship to Puerto Rico
Catalog No. 50-854-165 Supplier Bioanalytical Systems Inc Supplier No. CX2021S
Catheter, Short Femoral; Use to cannulate the femoral vein; Bead Color: Dark Blue; Dead Volume: 56uL; Each *Does not ship to Puerto Rico