Learn More
Bioanalytical Systems Inc CULEX MOUSE CAROTID-RED
SDP
Catalog No. 50854188
Encompass
Catheter, Mouse Carotid; Use to cannulate the carotid artery of a mouse; Bead Color: Red; Dead volume: 14.5uL; Each *Does not ship to Puerto Rico
Catalog No. 50-854-188 Supplier Bioanalytical Systems Inc Supplier No. CX2052S
Catheter, Mouse Carotid; Use to cannulate the carotid artery of a mouse; Bead Color: Red; Dead volume: 14.5uL; Each *Does not ship to Puerto Rico