Learn More
Bioanalytical Systems Inc CULEX MOUSE JUGULAR-LT BLUE
SDP
Catalog No. 50854169
Encompass
Catheter, Mouse Jugular; Use to cannulate the jugular vein of a mouse; Bead color: Light blue; Dead volume: 8uL; Each *Does not ship to Puerto Rico
Catalog No. 50-854-169 Supplier Bioanalytical Systems Inc Supplier No. CX2022S
Catheter, Mouse Jugular; Use to cannulate the jugular vein of a mouse; Bead color: Light blue; Dead volume: 8uL; Each *Does not ship to Puerto Rico