Learn More
Bioanalytical Systems Inc CULEX PLASTIC H2O TUBE-SS SIP
SDP
Supplier:  Bioanalytical Systems Inc CX5000
Encompass
Tube, Water; Culex; W/stainless steel sipper; For dual-species rat/mouse metabolic cage; Polycarbonate plastic; Autoclavable *Does not ship to Puerto Rico
Catalog No. 50-854-195