Learn More
Enzo Life Sciences Cullin-1 monoclonal antibody (AS97.1) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC135F
Encompass_Preferred
Clone: AS97.1. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human cullin 1. UniProt ID: Q13616. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Rabbit, Monkey, Bovine. Applications: WB. Recommended Dilutions: Western Blot (1:250, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.1mM PMSF and 50% glycerol. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7241