Learn More
Enzo Life Sciences Cullin-1 monoclonal antibody (AS97.1) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIKAMCC135D
Encompass_Preferred
Clone: AS97.1. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant human cullin 1. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat, Rabbit, Monkey, Bovine. Applications: WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.1mM PMSF and 50% glycerol.
Catalog No. 50-200-7240