Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cullin 4A/4B Antibody (YA2583) | 10 UL
SDP

Catalog No. 5000443581
Encompass_Preferred

Cullin 4A/4B Antibody (YA2583) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Cullin 4A/4B. It is supplied in PBS, pH 7.4, and is affinity purified. This antibody serves as the core component of multiple cullin-RING-based E3 ubiquitin-protein ligase complexes, facilitating ubiquitination and proteasomal degradation of target proteins, and plays roles in DNA repair/replication and cell cycle regulation.

  • Targets Cullin 4A/4B
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Affinity purified
  • Supports Western Blotting (WB) and Immunohistochemistry-Paraffin (IHC-P) applications
  • Involved in cell biology research
  • Available in liquid form
  • Stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles

Catalog No. 50-004-43581 Supplier Medchemexpress LLC Supplier No. HYP8283810UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cullin 4A/4B Antibody (YA2583) is a rabbit-derived, non-conjugated IgG monoclonal antibody that targets Cullin 4A/4B. It is supplied in PBS, pH 7.4, and is affinity purified. This antibody serves as the core component of multiple cullin-RING-based E3 ubiquitin-protein ligase complexes, facilitating ubiquitination and proteasomal degradation of target proteins, and plays roles in DNA repair/replication and cell cycle regulation.

  • Targets Cullin 4A/4B
  • Rabbit-derived and non-conjugated IgG monoclonal antibody
  • Affinity purified
  • Supports Western Blotting (WB) and Immunohistochemistry-Paraffin (IHC-P) applications
  • Involved in cell biology research
  • Available in liquid form
  • Stored at -20°C for 1 year, avoiding repeated freeze/thaw cycles

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.