Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cullin 4A/4B Antibody (YA2583) | 50 UL
SDP

Catalog No. 5000445292
Encompass_Preferred

Cullin 4A/4B Antibody (YA2583) is a Rabbit-derived, non-conjugated IgG monoclonal antibody, targeting Cullin 4A/4B. It is intended for research use only and is considered stable under recommended storage conditions.

  • Host: Rabbit
  • Reactivity: Human, Hamster, Rat
  • Clonality: YA2583
  • Swissprot ID: Q13620
  • Predicted band size: 104 kDa
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Concentration: 0.2 mg/mL

Catalog No. 50-004-45292 Supplier Medchemexpress LLC Supplier No. HYP8283850UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cullin 4A/4B Antibody (YA2583) is a Rabbit-derived, non-conjugated IgG monoclonal antibody, targeting Cullin 4A/4B. It is intended for research use only and is considered stable under recommended storage conditions.

  • Host: Rabbit
  • Reactivity: Human, Hamster, Rat
  • Clonality: YA2583
  • Swissprot ID: Q13620
  • Predicted band size: 104 kDa
  • Storage: Stored at -20°C for 1 year. Avoid repeated freeze/thaw cycles.
  • Concentration: 0.2 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.