Learn More
CELLTREAT Scientific Products CAP ONLY, 5mL Culture Tube Dual Cap, PE, Non-Sterile
SDP
Catalog No. NC2131760
Encompass_Preferred
Dual caps allow for either vented or sealed positions. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.
This item is not returnable. View return policy
Catalog No. NC2131760 Supplier CELLTREAT Scientific Products Supplier No. 230443
Dual caps allow for either vented or sealed positions. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.