Learn More
CELLTREAT Scientific Products TUBE ONLY, 5mL Culture Tube, PP, Non-Sterile
SDP
Supplier:  CELLTREAT Scientific Products 230431
Encompass_Preferred
Polypropylene tubes offer better chemical resistance. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.
Catalog No. NC2131767