Learn More
CELLTREAT Scientific Products TUBE ONLY, 5mL Culture Tube, PS, Non-Sterile
SDP
Supplier:  CELLTREAT Scientific Products 230432
Encompass_Preferred
Polystyrene tubes are optically clear to visualize cultures. Suitable for FACS and flow cytometry applications. Used for bacterial & suspension culture.
Catalog No. NC2131769