Learn More
Thermo Scientific™ Culti-Loops™ Achromobacter xylosoxidans subsp. xylosoxidans ATCC™ 27061™
Catalog No. R4601012 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 Loops/Pk.
Catalog No. Quantity
R4601012 5 Loops/Pk.
1 options
This item is not returnable. View return policy
Catalog No. R4601012 Supplier Thermo Scientific™ Supplier No. R4601012
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
Thermo Scientific™ Culti-Loops™ are ready-to-use QC organisms recommended for use in performance testing of media, stains, reagents and identification kits, and for the evaluation of bacteriological procedures.
The loop may be dissolved in liquid medium, Culti-Loops Aerobic Set Rehydration Fluid, Part No. R4631000 or streaked directly onto an appropriate media.
Ordering Information:
Culti-Loops are individually sealed in a foil pouch and packaged five loops per package.
TRUSTED_SUSTAINABILITY
Specifications
ATCC® 27061TM*
Certifications/Compliance Culti-Loops are CE Marked.
Description Achromobacter xylosoxidans subsp. Xylosoxidans
Product Type Culti-Loops Quality Control Organism
Target Organism Class Gram-negative Rods
Test Type Qualitative QC
Packaging Type 5 Loops/Pack
Quantity 5 Loops/Pk.
Strain Designations KM 543 [CCUG 12689, CIP 71.32, DSM 10346, DSM 2402, EY 543, GIFL 543, IFO 15126, JCM 9659, LMG 1863, NCTC 10807, NRRL B-4082, R. Hugh 2838]
*The ATCC Licensed Derivative Emblem, the ATCC Licensed Derivative word mark, and the ATCC catalog marks are trademarks of ATCC™ and are used under license.